COMH4211AD03 Combi-Blocks, Inc.

1-(Cyclopropylmethyl)-4-iodo-1h-pyrazole

  • MDL number MFCD20327530
  • Cas Number : 1340571-52-9
  • Molecular Weight : 248.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(cyclopropylmethyl)-4-iodo-1H-pyrazole
Inchi Code 1S/C7H9IN2/c8-7-3-9-10(5-7)4-6-1-2-6/h3,5-6H,1-2,4H2
Inchi Key SXBRZFVZNYEHIV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1897

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1897
Availability details for
Quantity
  • Availability
  • Add to Cart