COMH4211AD04 Combi-Blocks, Inc.

3-[2-(3,4-Difluoro-phenyl)-ethyl]-benzoic acid

  • MDL number MFCD31614031
  • Cas Number : 2270905-97-8
  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1898
Availability details for
Quantity
  • Availability
  • Add to Cart