COMH4211AD05 Combi-Blocks, Inc.

6-Bromo-1-cyclopentyl-1h-indole-3-carbonitrile

  • MDL number MFCD31583010
  • Cas Number : 1510992-26-3
  • Molecular Weight : 289.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1899
Availability details for
Quantity
  • Availability
  • Add to Cart