COMH4211AD06 Combi-Blocks, Inc.

S -N-(1-Phenylethyl)-isophthalamic acid

  • MDL number MFCD30830168
  • Cas Number : 169690-59-9
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1901

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1901
Availability details for
Quantity
  • Availability
  • Add to Cart