COMH4211AD08 Combi-Blocks, Inc.

1-(Bromomethyl)-2,5-difluoro-4-methylbenzene

  • MDL number MFCD23838739
  • Cas Number : 1515657-97-2
  • Molecular Weight : 221.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1903

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1903
Availability details for
Quantity
  • Availability
  • Add to Cart