COMH4211AD0B Combi-Blocks, Inc.

Cis 4-tert-butoxycarbonylamino-cyclohexanecarboxylic acid 2,5-dioxo-pyrrolidin-1-yl ester

  • MDL number MFCD29917459
  • Cas Number : 271577-34-5
  • Molecular Weight : 340.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1909

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1909
Availability details for
Quantity
  • Availability
  • Add to Cart