COMH4211AD0C Combi-Blocks, Inc.

5-Bromo-2-(1,1-difluoroethyl)pyrimidine

  • MDL number MFCD30003554
  • Cas Number : 1211530-92-5
  • Molecular Weight : 223.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1910
Availability details for
Quantity
  • Availability
  • Add to Cart