COMH4211AD0D Combi-Blocks, Inc.

1-(2-Methyl-benzyl)-1h-pyrazol-4-ol

  • MDL number MFCD31561483
  • Cas Number : 1603517-86-7
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1917
Availability details for
Quantity
  • Availability
  • Add to Cart