COMH4211AD0E Combi-Blocks, Inc.

Pentanedioic acid, 1-(2,5-dioxo-1-pyrrolidinyl) 5-(2-propyn-1-yl) ester

  • MDL number MFCD31381641
  • Cas Number : 1446429-04-4
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1921

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1921
Availability details for
Quantity
  • Availability
  • Add to Cart