COMH4211AD0F Combi-Blocks, Inc.

2-Bromo-n-cyclobutyl-5-methylaniline

  • MDL number MFCD24301684
  • Cas Number : 1548566-83-1
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1924

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1924
Availability details for
Quantity
  • Availability
  • Add to Cart