COMH4211AD10 Combi-Blocks, Inc.

7-(3-((2-Aminoethyl)amino)-3-oxopropyl)-5,5-difluoro-1,3-dimethyl-5h-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide

  • MDL number MFCD28400780
  • Cas Number : 220524-70-9
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1928

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1928
Availability details for
Quantity
  • Availability
  • Add to Cart