COMH4211AD11 Combi-Blocks, Inc.

2-(4-Isobutoxy-pyrazol-1-yl)-phenylamine

  • MDL number MFCD31579543
  • Cas Number : 2270908-72-8
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1930

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1930
Availability details for
Quantity
  • Availability
  • Add to Cart