COMH4211AD12 Combi-Blocks, Inc.

4-(2,2-Difluoroethoxy)phenol

  • MDL number MFCD12807647
  • Cas Number : 1183003-98-6
  • Molecular Weight : 174.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1931
Availability details for
Quantity
  • Availability
  • Add to Cart