COMH4211AD15 Combi-Blocks, Inc.

2',4'-Dichloro-[1,1'-biphenyl]-4-amine

  • MDL number MFCD09026457
  • Cas Number : 939757-52-5
  • Molecular Weight : 238.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1936

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1936
Availability details for
Quantity
  • Availability
  • Add to Cart