COMH4211AD16 Combi-Blocks, Inc.

2-(Difluoromethoxy)-n-[4-(2-methyl-1,3-oxazol-5-yl)phenyl]benzamide

  • MDL number MFCD22196910
  • Cas Number : 1311855-96-5
  • Molecular Weight : 344.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1938

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1938
Availability details for
Quantity
  • Availability
  • Add to Cart