COMH4211AD17 Combi-Blocks, Inc.

5-Bromo-n1-(4-methoxyphenyl)benzene-1,2-diamine

  • MDL number MFCD21513450
  • Cas Number : 1416336-77-0
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1939

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1939
Availability details for
Quantity
  • Availability
  • Add to Cart