COMH4211AD18 Combi-Blocks, Inc.

(5-Amino-2-methyl-phenyl)-pyrrolidin-1-yl-methanone

  • MDL number MFCD14618159
  • Cas Number : 1249437-45-3
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1943

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1943
Availability details for
Quantity
  • Availability
  • Add to Cart