COMH4211AD19 Combi-Blocks, Inc.

(S)-2-(4-Trifluoromethoxyphenoxymethyl)-pyrrolidine

  • MDL number MFCD11849593
  • Cas Number : 681482-14-4
  • Molecular Weight : 261.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1944

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1944
Availability details for
Quantity
  • Availability
  • Add to Cart