COMH4211AD1A Combi-Blocks, Inc.

4-Amino-n,n-dimethyl-2-(trifluoromethyl)benzamide

  • MDL number MFCD29097138
  • Cas Number : 1174234-00-4
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1948

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1948
Availability details for
Quantity
  • Availability
  • Add to Cart