COMH4211AD1C Combi-Blocks, Inc.

4-Bromo-n,n-diethyl-2-methylbenzamide

  • MDL number MFCD20543578
  • Cas Number : 682778-12-7
  • Molecular Weight : 270.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1952
Availability details for
Quantity
  • Availability
  • Add to Cart