COMH4211AD1D Combi-Blocks, Inc.

4-Bromo-2-(4-methoxy-benzyloxy)-1-nitro-benzene

  • MDL number MFCD31613860
  • Cas Number : 1628677-57-5
  • Molecular Weight : 338.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1954

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1954
Availability details for
Quantity
  • Availability
  • Add to Cart