COMH4211AD20 Combi-Blocks, Inc.

N-(2,2-Dimethoxyethyl)maleamic acid

  • MDL number MFCD31381662
  • Cas Number : 76619-99-3
  • Molecular Weight : 203.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (Z)-4-((2,2-dimethoxyethyl)amino)-4-oxobut-2-enoic acid
Inchi Code 1S/C8H13NO5/c1-13-8(14-2)5-9-6(10)3-4-7(11)12/h3-4,8H,5H2,1-2H3,(H,9,10)(H,11,12)/b4-3-
Inchi Key VACQOXIVQVPQKN-ARJAWSKDSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1959
Availability details for
Quantity
  • Availability
  • Add to Cart