COMH4211AD23 Combi-Blocks, Inc.

3-(Propylcarbamoyl)propanoic acid

  • MDL number MFCD01098172
  • Cas Number : 61283-60-1
  • Molecular Weight : 159.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1967

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1967
Availability details for
Quantity
  • Availability
  • Add to Cart