COMH4211AD24 Combi-Blocks, Inc.

1-(3-Fluoro-4-methyl-benzyl)-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD30830182
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1968

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1968
Availability details for
Quantity
  • Availability
  • Add to Cart