COMH4211AD27 Combi-Blocks, Inc.

Benzamide, n-(5-bromo-2-pyridinyl)-4-methyl-

  • MDL number MFCD00984047
  • Cas Number : 86843-83-6
  • Molecular Weight : 291.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1972

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1972
Availability details for
Quantity
  • Availability
  • Add to Cart