COMH4211AD28 Combi-Blocks, Inc.

Methyl 2-(cyclobutylamino)-5-fluorobenzoate

  • MDL number MFCD16108410
  • Cas Number : 1249421-82-6
  • Molecular Weight : 223.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1974

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1974
Availability details for
Quantity
  • Availability
  • Add to Cart