COMH4211AD29 Combi-Blocks, Inc.

3-Methyl-4-pyridin-2-ylisoxazolo[3,4-d]pyridazin-7(6h)-one

  • MDL number MFCD31561592
  • Cas Number : 720718-35-4
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1975

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1975
Availability details for
Quantity
  • Availability
  • Add to Cart