COMH4211AD2A Combi-Blocks, Inc.

1-Ethyl-5-fluoro-1h-indole-2-carboxylic acid

  • MDL number MFCD23161718
  • Cas Number : 1391467-29-0
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1977

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1977
Availability details for
Quantity
  • Availability
  • Add to Cart