COMH4211AD2D Combi-Blocks, Inc.

1H-Pyrazole, 4-iodo-1-[[2-(trimethylsilyl)ethoxy]methyl]-

  • MDL number MFCD20483624
  • Cas Number : 220299-49-0
  • Molecular Weight : 324.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1982

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1982
Availability details for
Quantity
  • Availability
  • Add to Cart