COMH4211AD2F Combi-Blocks, Inc.

4-Bromo-2-cyclopropylmethoxy-phenylamine

  • MDL number MFCD21193997
  • Cas Number : 1369781-99-6
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1985

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1985
Availability details for
Quantity
  • Availability
  • Add to Cart