COMH4211AD30 Combi-Blocks, Inc.

3-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazol-1-ylmethyl]-benzoic acid methyl ester

  • MDL number MFCD21337501
  • Cas Number : 1404431-51-1
  • Molecular Weight : 342.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1987

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1987
Availability details for
Quantity
  • Availability
  • Add to Cart