COMH4211AD31 Combi-Blocks, Inc.

N-[(3-Bromo-4-methoxyphenyl)methyl]cyclopropanamine

  • MDL number MFCD12146172
  • Cas Number : 1152928-07-8
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1988

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1988
Availability details for
Quantity
  • Availability
  • Add to Cart