COMH4211AD32 Combi-Blocks, Inc.

3-(4-Methoxyphenoxy)azetidine

  • MDL number MFCD09756536
  • Cas Number : 954220-70-3
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1989

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1989
Availability details for
Quantity
  • Availability
  • Add to Cart