COMH4211AD34 Combi-Blocks, Inc.

1-Methanesulfonyl-4-methylpiperidin-4-amine

  • MDL number MFCD20721560
  • Cas Number : 651056-95-0
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1994

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1994
Availability details for
Quantity
  • Availability
  • Add to Cart