COMH4211AD36 Combi-Blocks, Inc.

1-[(5-Bromopyridin-2-yl)carbonyl]piperazine

  • MDL number MFCD17259126
  • Cas Number : 1284188-09-5
  • Molecular Weight : 270.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1996
Availability details for
Quantity
  • Availability
  • Add to Cart