COMH4211AD37 Combi-Blocks, Inc.

1-Benzyl-3-(4-bromophenyl)urea

  • MDL number MFCD00017794
  • Cas Number : 13208-62-3
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1999

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1999
Availability details for
Quantity
  • Availability
  • Add to Cart