COMH4211AD38 Combi-Blocks, Inc.

1-[3-(Cyclopropylmethoxy)phenyl]ethanone

  • MDL number MFCD12138820
  • Cas Number : 478163-32-5
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2001
Availability details for
Quantity
  • Availability
  • Add to Cart