COMH4211AD39 Combi-Blocks, Inc.

1-(Ethylsulfonyl)-4-piperidineethanol

  • MDL number MFCD28127467
  • Cas Number : 1435489-63-6
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2006

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2006
Availability details for
Quantity
  • Availability
  • Add to Cart