COMH4211AD3A Combi-Blocks, Inc.

2-(2-Fluoro-4-methoxy-benzylidene)-malononitrile

  • MDL number MFCD06214099
  • Cas Number : 582306-88-5
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2011
Availability details for
Quantity
  • Availability
  • Add to Cart