COMH4211AD3B Combi-Blocks, Inc.

2-Ethoxy-4-nitrobenzonitrile

  • MDL number MFCD11042855
  • Cas Number : 642081-59-2
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2012

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2012
Availability details for
Quantity
  • Availability
  • Add to Cart