COMH4211AD3C Combi-Blocks, Inc.

(2-Allyloxy-ethoxy)-acetic acid benzyl ester

  • MDL number MFCD31583127
  • Cas Number : 909570-35-0
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2014
Availability details for
Quantity
  • Availability
  • Add to Cart