COMH4211AD3E Combi-Blocks, Inc.

1-(5-Bromo-2-nitro-phenyl)-1h-pyrazol-4-ol

  • MDL number MFCD31579302
  • Cas Number : 2270905-29-6
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2018

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2018
Availability details for
Quantity
  • Availability
  • Add to Cart