COMH4211AD41 Combi-Blocks, Inc.

Ethyl[(4-fluoro-2-methylphenyl)methyl]amine

  • MDL number MFCD16686578
  • Cas Number : 1248091-26-0
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2023
Availability details for
Quantity
  • Availability
  • Add to Cart