COMH4211AD42 Combi-Blocks, Inc.

[4-Methyl-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridin-2-yl]-morpholin-4-yl-methanone

  • MDL number MFCD31613900
  • Molecular Weight : 332.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2028
Availability details for
Quantity
  • Availability
  • Add to Cart