COMH4211AD43 Combi-Blocks, Inc.

2,5-Dioxopyrrolidin-1-yl 4,4-difluorocyclohexanecarboxylate

  • MDL number MFCD27964349
  • Cas Number : 1369515-34-3
  • Molecular Weight : 261.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2029
Availability details for
Quantity
  • Availability
  • Add to Cart