COMH4211AD44 Combi-Blocks, Inc.

2-(3,3-Dimethyl-piperidin-4-yl)-ethanol hydrochloride

  • MDL number MFCD31579412
  • Cas Number : 2270907-12-3
  • Molecular Weight : 193.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2030
Availability details for
Quantity
  • Availability
  • Add to Cart