COMH4211AD46 Combi-Blocks, Inc.

(3-Fluoro-5-methoxy-4-nitrophenyl)methanol

  • MDL number MFCD20527929
  • Cas Number : 1257997-29-7
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2034

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2034
Availability details for
Quantity
  • Availability
  • Add to Cart