COMH4211AD47 Combi-Blocks, Inc.

3-Bromobenzyl 4-nitrophenyl carbonate

  • MDL number MFCD31583354
  • Cas Number : 1404431-58-8
  • Molecular Weight : 352.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2035

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2035
Availability details for
Quantity
  • Availability
  • Add to Cart