COMH4211AD4C Combi-Blocks, Inc.

2-(4-Bromo-2-fluoro-phenyl)-1-piperidin-1-yl-ethanone

  • MDL number MFCD31613958
  • Cas Number : 2270906-15-3
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2045
Availability details for
Quantity
  • Availability
  • Add to Cart