COMH4211AD4D Combi-Blocks, Inc.

6-Chloro-n-ethyl-n'-propyl-1,3,5-triazine-2,4-diamine

  • MDL number MFCD00466645
  • Cas Number : 90952-64-0
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2047

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2047
Availability details for
Quantity
  • Availability
  • Add to Cart